4-((benzyloxy)methyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

125233-1G 125233-500MG 125233-5G 125233-25G
77464-406EA 415.09 USD
77464-406 77477-194 77487-286 77498-100
4-((benzyloxy)methyl)benzaldehyde ≥95%
4-((benzyloxy)methyl)benzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
CAS Number: 216384-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR