(4-((benzyloxy)methyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

125228-1G 125228-500MG 125228-5G 125228-25G
77464-402EA 258.53 USD
77464-402 77477-190 77487-282 77498-096
(4-((benzyloxy)methyl)phenyl)boronic acid ≥95%
(4-((benzyloxy)methyl)phenyl)boronic acid

Precaución: For research use only

Formula: C₁₄H₁₅BO₃
Storage Temperature: Ambient
CAS Number: 163680-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR