8-(2,4-Difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

124866-1G 124866-500MG 124866-5G 124866-25G
77464-398EA 397.58 USD
77464-398 77477-186 77487-278 77498-090
8-(2,4-Difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(2,4-Difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₄H₁₆F₂O₃
MW: 270.2718 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20862285
CAS Number: 1493364-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR