tert-Butyl 4-hydroxy-4-(4-(trifluoromethyl)phenyl)piperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

124776-1G 124776-500MG 124776-5G
77464-390EA 444.96 USD
77464-390 77477-178 77487-270
tert-Butyl 4-hydroxy-4-(4-(trifluoromethyl)phenyl)piperidine-1-carboxylate ≥95%
tert-Butyl 4-hydroxy-4-(4-(trifluoromethyl)phenyl)piperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₇H₂₂F₃NO₃
MW: 345.3566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20039766
CAS Number: 1191240-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR