tert-Butyl 4-(2-fluorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

124768-1G 124768-500MG 124768-5G 124768-25G
77464-386EA 147.29 USD
77464-386 77477-174 77487-266 77498-082
tert-Butyl 4-(2-fluorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl 4-(2-fluorophenyl)-4-hydroxypiperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₆H₂₂FNO₃
MW: 295.3491 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953393
CAS Number: 403806-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR