1-(2-Bromo-5-fluoropyridin-4-yl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

124638-1G 124638-500MG 124638-5G 124638-25G
77464-378EA 415.09 USD
77464-378 77477-166 77487-258 77498-078
1-(2-Bromo-5-fluoropyridin-4-yl)-2-chloro-2,2-difluoroethanone ≥95%
1-(2-Bromo-5-fluoropyridin-4-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₂BrClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD34784710
CAS Number: 2918895-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR