2-Fluoro-1-(2,3,5-trichloropyridin-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

124614-1G 124614-500MG 124614-5G 124614-25G
77464-374EA 415.09 USD
77464-374 77477-162 77487-254 77498-074
2-Fluoro-1-(2,3,5-trichloropyridin-4-yl)ethanone ≥97%
2-Fluoro-1-(2,3,5-trichloropyridin-4-yl)ethanone

Precaución: For research use only

Formula: C₇H₃Cl₃FNO
MW: 242.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825908
CAS Number: 2969397-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR