1-(2-Bromo-5-chloropyridin-4-yl)-2-fluoroethanone ≥97%

Supplier: AOBChem USA

Warning

124595-1G 124595-500MG 124595-5G 124595-25G
77464-370EA 415.09 USD
77464-370 77477-160 77487-250 77498-070
1-(2-Bromo-5-chloropyridin-4-yl)-2-fluoroethanone ≥97%
1-(2-Bromo-5-chloropyridin-4-yl)-2-fluoroethanone

Precaución: For research use only

Formula: C₇H₄BrClFNO
MW: 252.468 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825902
CAS Number: 2969219-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR