1-(3-Chloro-2-methoxypyridin-4-yl)cyclopentanol ≥96%

Supplier: AOBChem USA

Warning

124568-1G 124568-500MG 124568-5G 124568-25G
77464-366EA 312.09 USD
77464-366 77477-156 77487-246 77498-066
1-(3-Chloro-2-methoxypyridin-4-yl)cyclopentanol ≥96%
1-(3-Chloro-2-methoxypyridin-4-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₁H₁₄ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34825893
CAS Number: 2150933-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR