1-(2-Chloropyridin-3-yl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

124520-1G 124520-500MG 124520-5G
77464-362EA 715.85 USD
77464-362 77477-152 77487-242
1-(2-Chloropyridin-3-yl)-2-fluoroethanone ≥95%
1-(2-Chloropyridin-3-yl)-2-fluoroethanone

Precaución: For research use only

Formula: C₇H₅ClFNO
MW: 173.572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19689732
CAS Number: 1234552-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR