Benzo[d][1,3]dioxol-4-ylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Methylenedioxyphenylboronic acid

12451-1G 12451-500MG 12451-5G 12451-25G
77464-358EA 391.4 USD
77464-358 77477-150 77487-240 77498-060
Benzo[d][1,3]dioxol-4-ylboronic acid ≥95%
Benzo[d][1,3]dioxol-4-ylboronic acid

Precaución: For research use only

Formula: C₇H₇BO₄
MW: 165.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095113
CAS Number: 361456-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR