Methyl 3-iodo-4-Methyl-5-nitrobenzoate ≥95%

Supplier: AOBChem USA

Warning

124058-1G 124058-500MG 124058-5G
77464-350EA 752.93 USD
77464-350 77477-140 77487-230
Methyl 3-iodo-4-Methyl-5-nitrobenzoate ≥95%
Methyl 3-iodo-4-Methyl-5-nitrobenzoate

Precaución: For research use only

Formula: C₉H₈INO₄
Storage Temperature: Ambient
MDL Number: MFCD07781360
CAS Number: 21323-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR