1-(3-Bromo-2,6-difluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

123504-1G 123504-500MG 123504-5G
77464-342EA 597.4 USD
77464-342 77477-132 77487-220
1-(3-Bromo-2,6-difluorophenyl)cyclobutanol ≥95%
1-(3-Bromo-2,6-difluorophenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₀H₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34825411
CAS Number: 1699022-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR