(3-Ethoxy-2-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

122944-1G 122944-500MG 122944-5G 122944-25G
77464-338EA 415.09 USD
77464-338 77477-128 77487-216 77498-034
(3-Ethoxy-2-methylphenyl)(methyl)sulfane ≥95%
(3-Ethoxy-2-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34825131
CAS Number: 2918960-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR