(3-Ethoxy-2-methylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

122938-1G 122938-500MG 122938-5G
77464-334EA 572.68 USD
77464-334 77477-124 77487-212
(3-Ethoxy-2-methylphenyl)boronic acid ≥95%
(3-Ethoxy-2-methylphenyl)boronic acid

Precaución: For research use only

Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD28009351
CAS Number: 1383575-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR