1-(6-Bromo-2-fluoro-3-(methylsulfonyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

122173-1G 122173-500MG 122173-5G 122173-25G
77464-330EA 214.24 USD
77464-330 77477-122 77487-208 77498-022
1-(6-Bromo-2-fluoro-3-(methylsulfonyl)phenyl)ethanol ≥95%
1-(6-Bromo-2-fluoro-3-(methylsulfonyl)phenyl)ethanol

Precaución: For research use only

Formula: C₉H₁₀BrFO₃S
MW: 297.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34824789
CAS Number: 2969437-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR