2,2-Difluoro-5-(methylthio)benzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

121834-1G 121834-500MG 121834-5G 121834-25G
77464-326EA 415.09 USD
77464-326 77477-118 77487-204 77498-016
2,2-Difluoro-5-(methylthio)benzo[d][1,3]dioxole ≥95%
2,2-Difluoro-5-(methylthio)benzo[d][1,3]dioxole

Precaución: For research use only

Formula: C₈H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD31923561
CAS Number: 2565806-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR