(iodoethynyl)benzene ≥97%

Supplier: AOBChem USA

Warning

121533-1G 121533-500MG 121533-5G 121533-25G
77464-322EA 112.27 USD
77464-322 77477-114 77487-200 77498-012
(iodoethynyl)benzene ≥97%
(iodoethynyl)benzene

Precaución: For research use only

Formula: C₈H₅I
Storage Temperature: Ambient
MDL Number: MFCD00457145
CAS Number: 932-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR