2-(methylthio)-6-(trimethylsilyl)phenol ≥97%

Supplier: AOBChem USA

Warning

120912-1G 120912-500MG 120912-5G 120912-25G
77464-318EA 245.14 USD
77464-318 77477-110 77487-196 77498-006
2-(methylthio)-6-(trimethylsilyl)phenol ≥97%
2-(methylthio)-6-(trimethylsilyl)phenol

Precaución: For research use only

Formula: C₁₀H₁₆OSSi
MW: 212.384 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34824253
CAS Number: 2918960-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR