2-formyl-4-methoxyphenyl acetate ≥97%

Supplier: AOBChem USA

Warning

120890-1G 120890-500MG 120890-5G
77464-314EA 685.98 USD
77464-314 77477-106 77487-192
2-formyl-4-methoxyphenyl acetate ≥97%
2-formyl-4-methoxyphenyl acetate

Precaución: For research use only

Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00599106
CAS Number: 62536-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR