1-Bromo-2-fluoro-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

120637-1G 120637-500MG 120637-5G
77464-310EA 483.07 USD
77464-310 77477-102 77487-186
1-Bromo-2-fluoro-3-(methylsulfonyl)benzene ≥97%
1-Bromo-2-fluoro-3-(methylsulfonyl)benzene

Precaución: For research use only

Formula: C₇H₆BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD26687709
CAS Number: 1541305-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR