1-(4-(Methylthio)phenyl)propan-1-one ≥96%

Supplier: AOBChem USA

Warning

120602-1G 120602-500MG 120602-5G 120602-25G
77464-298EA 309 USD
77464-298 77477-090 77487-174 77497-994
1-(4-(Methylthio)phenyl)propan-1-one ≥96%
1-(4-(Methylthio)phenyl)propan-1-one

Precaución: For research use only

Formula: C₁₀H₁₂OS
MW: 180.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863479
CAS Number: 52129-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR