Ethyl 2-(4-ethoxy-2,3-difluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

120384-1G 120384-500MG 120384-5G
77464-294EA 666.41 USD
77464-294 77477-086 77487-170
Ethyl 2-(4-ethoxy-2,3-difluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-ethoxy-2,3-difluorophenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₂H₁₂F₂O₄
MW: 258.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34824058
CAS Number: 1898399-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR