1-(4-Ethoxy-2,3-difluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

120381-1G 120381-500MG 120381-5G 120381-25G
77464-290EA 415.09 USD
77464-290 77477-082 77487-166 77497-988
1-(4-Ethoxy-2,3-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-Ethoxy-2,3-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₀H₇F₅O₂
MW: 254.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34824055
CAS Number: 2969465-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR