2-Methyl-2-(3-(methylthio)thiophen-2-yl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

119859-1G 119859-500MG 119859-5G 119859-25G
77464-286EA 126.69 USD
77464-286 77477-078 77487-162 77497-984
2-Methyl-2-(3-(methylthio)thiophen-2-yl)-1,3-dioxolane ≥97%
2-Methyl-2-(3-(methylthio)thiophen-2-yl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₁₂O₂S₂
MW: 216.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823902
CAS Number: 2918960-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR