2-acetylthiophene-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

119856-1G 119856-500MG 119856-5G 119856-25G
77464-282EA 250.29 USD
77464-282 77477-074 77487-158 77497-980
2-acetylthiophene-3-carboxylic acid ≥97%
2-acetylthiophene-3-carboxylic acid

Precaución: For research use only

Formula: C₇H₆O₃S
Storage Temperature: Ambient
MDL Number: MFCD00159556
CAS Number: 30006-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR