5-Bromo-8-fluoro-7-iodoquinoline ≥97%

Supplier: AOBChem USA

Warning

119837-1G 119837-500MG 119837-5G 119837-25G
77464-278EA 415.09 USD
77464-278 77477-070 77487-154 77497-976
5-Bromo-8-fluoro-7-iodoquinoline ≥97%
5-Bromo-8-fluoro-7-iodoquinoline

Precaución: For research use only

Formula: C₉H₄BrFIN
Storage Temperature: Ambient
MDL Number: MFCD34823892
CAS Number: 2918963-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR