5-Bromo-8-fluoroquinoline-7-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

119832-1G 119832-500MG 119832-5G 119832-25G
77464-274EA 415.09 USD
77464-274 77477-066 77487-150 77497-972
5-Bromo-8-fluoroquinoline-7-carboxylic acid ≥97%
5-Bromo-8-fluoroquinoline-7-carboxylic acid

Precaución: For research use only

Formula: C₁₀H₅BrFNO₂
MW: 270.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823888
CAS Number: 2918947-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR