Methyl(naphthalen-1-yl)sulfane ≥96%

Supplier: AOBChem USA

Warning

119683-1G 119683-500MG 119683-5G
77464-270EA 648.9 USD
77464-270 77477-062 77487-146
Methyl(naphthalen-1-yl)sulfane ≥96%
Methyl(naphthalen-1-yl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₀S
Storage Temperature: Ambient
MDL Number: MFCD11656009
CAS Number: 10075-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR