5-(methylthio)isoquinoline ≥97%
Supplier: AOBChem USA
|
Warning
|
119647-1G
119647-500MG
119647-5G
77464-266EA
1335.91
USD
77464-266
77477-058
77487-142
5-(methylthio)isoquinoline ≥97%
5-(methylthio)isoquinoline
Precaución: For research use only
Formula:
C₁₀H₉NS MW: 175.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28668214 CAS Number: 90265-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...