2-Bromo-4-chloro-5-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

119485-1G 119485-500MG 119485-5G 119485-25G
77464-262EA 93.73 USD
77464-262 77477-054 77487-138 77497-968
2-Bromo-4-chloro-5-fluoroaniline ≥97%
2-Bromo-4-chloro-5-fluoroaniline

Precaución: For research use only

Formula: C₆H₄BrClFN
Storage Temperature: Ambient
MDL Number: MFCD12026231
CAS Number: 120694-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR