2-Chloro-6-methoxy-3-methylbenzoic acid ≥96%

Supplier: AOBChem USA

Warning

119351-1G 119351-500MG 119351-5G
77464-258EA 546.93 USD
77464-258 77477-050 77487-134
2-Chloro-6-methoxy-3-methylbenzoic acid ≥96%
2-Chloro-6-methoxy-3-methylbenzoic acid

Precaución: For research use only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823779
CAS Number: 1784293-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR