2-Chloro-6-hydroxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

119335-1G 119335-500MG 119335-5G 119335-25G
77464-254EA 392.43 USD
77464-254 77477-046 77487-130 77497-964
2-Chloro-6-hydroxy-3-methylbenzoic acid ≥97%
2-Chloro-6-hydroxy-3-methylbenzoic acid

Precaución: For research use only

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823765
CAS Number: 1781625-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR