2-(1,1-Difluoroethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

119110-1G 119110-500MG 119110-5G
77464-242EA 733.36 USD
77464-242 77477-034 77487-118
2-(1,1-Difluoroethyl)benzoic acid ≥95%
2-(1,1-Difluoroethyl)benzoic acid

Precaución: For research use only

Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20659035
CAS Number: 892390-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR