(4-((difluoromethyl)thio)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

119016-1G 119016-500MG 119016-5G
77464-238EA 622.12 USD
77464-238 77477-030 77487-114
(4-((difluoromethyl)thio)phenyl)boronic acid ≥97%
(4-((difluoromethyl)thio)phenyl)boronic acid

Precaución: For research use only

Formula: C₇H₇BF₂O₂S
MW: 204.002 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11183311
CAS Number: 1313761-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR