2,6-Dimethoxy-4-formylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (4-Formyl-2,6-dimethoxyphenyl)boronic acid

11880-1G 11880-500MG 11880-5G 11880-25G
77464-234EA 302.82 USD
77464-234 77477-026 77487-108 77497-950
2,6-Dimethoxy-4-formylphenylboronic acid ≥95%
2,6-Dimethoxy-4-formylphenylboronic acid

Precaución: For research use only

Formula: C₉H₁₁BO₅
MW: 209.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039020
CAS Number: 1256355-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR