Naphthalen-2-yl(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118558-1G 118558-500MG 118558-5G 118558-25G
77464-230EA 74.16 USD
77464-230 77477-022 77487-104 77497-946
Naphthalen-2-yl(phenyl)methanol ≥95%
Naphthalen-2-yl(phenyl)methanol

Precaución: For research use only

Formula: C₁₇H₁₄O
MW: 234.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01313901
CAS Number: 35060-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR