(4-Methoxy-3,5-dimethylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118551-1G 118551-500MG 118551-5G 118551-25G
77464-226EA 415.09 USD
77464-226 77477-018 77487-100 77497-944
(4-Methoxy-3,5-dimethylphenyl)(phenyl)methanol ≥95%
(4-Methoxy-3,5-dimethylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD25866147
CAS Number: 840-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR