(3-Methoxy-4-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118507-1G 118507-500MG 118507-5G 118507-25G
77464-222EA 253.38 USD
77464-222 77477-014 77487-096 77497-940
(3-Methoxy-4-methylphenyl)(phenyl)methanol ≥95%
(3-Methoxy-4-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16356979
CAS Number: 37825-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR