(3-Fluoro-5-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118482-1G 118482-500MG 118482-5G 118482-25G
77464-218EA 415.09 USD
77464-218 77477-010 77487-092 77497-936
(3-Fluoro-5-methylphenyl)(phenyl)methanol ≥95%
(3-Fluoro-5-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₃FO
MW: 216.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23750744
CAS Number: 1517000-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR