(4-Butylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118472-1G 118472-500MG 118472-5G 118472-25G
77464-214EA 341.96 USD
77464-214 77477-006 77487-088 77497-932
(4-Butylphenyl)(phenyl)methanol ≥95%
(4-Butylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₇H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD11096259
CAS Number: 944515-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR