(4-cyclohexylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118469-1G 118469-500MG 118469-5G 118469-25G
77464-210EA 415.09 USD
77464-210 77477-002 77487-084 77497-928
(4-cyclohexylphenyl)(phenyl)methanol ≥95%
(4-cyclohexylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₉H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD34784111
CAS Number: 72396-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR