(4-Ethoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118463-1G 118463-500MG 118463-5G 118463-25G
77464-206EA 415.09 USD
77464-206 77476-998 77487-080 77497-924
(4-Ethoxyphenyl)(phenyl)methanol ≥95%
(4-Ethoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00484071
CAS Number: 34979-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR