4-(Methylthio)benzhydrol ≥97%

Supplier: AOBChem USA

Warning

118460-1G 118460-500MG 118460-5G 118460-25G
77464-202EA 302.82 USD
77464-202 77476-994 77487-076 77497-920
4-(Methylthio)benzhydrol ≥97%
4-(Methylthio)benzhydrol

Precaución: For research use only

Formula: C₁₄H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD00093026
CAS Number: 183729-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR