(3-(Benzyloxy)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118447-1G 118447-500MG 118447-5G 118447-25G
77464-198EA 397.58 USD
77464-198 77476-990 77487-072 77497-914
(3-(Benzyloxy)phenyl)(phenyl)methanol ≥95%
(3-(Benzyloxy)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₂₀H₁₈O₂
MW: 290.356 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784106
CAS Number: 286947-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR