(3-(Methylthio)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118443-1G 118443-500MG 118443-5G 118443-25G
77464-194EA 415.09 USD
77464-194 77476-986 77487-068 77497-910
(3-(Methylthio)phenyl)(phenyl)methanol ≥95%
(3-(Methylthio)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₄OS
MW: 230.325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784105
CAS Number: 2918904-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR