(3-(tert-Butyl)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118439-1G 118439-500MG 118439-5G 118439-25G
77464-190EA 415.09 USD
77464-190 77476-982 77487-064 77497-906
(3-(tert-Butyl)phenyl)(phenyl)methanol ≥95%
(3-(tert-Butyl)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₇H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD34784102
CAS Number: 52458-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR