phenyl(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118437-1G 118437-500MG 118437-5G 118437-25G
77464-186EA 221.45 USD
77464-186 77476-978 77487-060 77497-902
phenyl(m-tolyl)methanol ≥95%
phenyl(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD01652338
CAS Number: 21945-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR