(2-Isopropoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118429-1G 118429-500MG 118429-5G 118429-25G
77464-182EA 370.8 USD
77464-182 77476-974 77487-056 77497-898
(2-Isopropoxyphenyl)(phenyl)methanol ≥95%
(2-Isopropoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
MW: 242.313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22950737
CAS Number: 93014-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR