(2-Isopropylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118424-1G 118424-500MG 118424-5G 118424-25G
77464-178EA 397.58 USD
77464-178 77476-970 77487-052 77497-894
(2-Isopropylphenyl)(phenyl)methanol ≥95%
(2-Isopropylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD34784098
CAS Number: 21945-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR